missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ 3-(tert-Butyldimethylsiloxy)glutaric anhydride, 98%

Manufacturer:  Alfa Aesar™ H6122714

 View more versions of this product

Catalog No. AAH6122714



Specifications

Specifications

3-(tert-Butyldimethylsiloxy)glutaric anhydride
1.4510
79°C to 81°C
91424-40-7
MFCD00075235
RXAJGRHLLRGVSB-UHFFFAOYSA-N
4-[tert-butyl(dimethyl)silyl]oxyoxane-2,6-dione
4141040
Solid
1.030g/mL
Moisture Sensitive
25g
C11H20O4Si
3-tert-butyldimethylsilyloxy glutaric anhydride, 4-tert-butyldimethylsilyl oxy oxane-2,6-dione, 3-tert-butyldimethylsilyl oxy pentanedioic anhydride, 3-tert-butyldimethylsilyloxy glutaric anhydride,, 4-tert-butyldimethylsilyl oxy dihydro-2h-pyran-2,6 3h-dione, acmc-209rbg, 4-tert-butyl dimethyl silyl oxyoxane-2,6-dione, 3-tert-butyldimethylsilyloxy glutaricanhydride, 3-tert-butyldimethylsiloxy pentanedioic anhydride, 3-tert-butyldimethyl silyl oxy glutaric anhydride
CC(C)(C)[Si](C)(C)OC1CC(=O)OC(=O)C1
244.362
244.36
98%

Chemical Identifiers

91424-40-7
244.362
RXAJGRHLLRGVSB-UHFFFAOYSA-N
4141040
CC(C)(C)[Si](C)(C)OC1CC(=O)OC(=O)C1
C11H20O4Si
MFCD00075235
3-tert-butyldimethylsilyloxy glutaric anhydride, 4-tert-butyldimethylsilyl oxy oxane-2,6-dione, 3-tert-butyldimethylsilyl oxy pentanedioic anhydride, 3-tert-butyldimethylsilyloxy glutaric anhydride,, 4-tert-butyldimethylsilyl oxy dihydro-2h-pyran-2,6 3h-dione, acmc-209rbg, 4-tert-butyl dimethyl silyl oxyoxane-2,6-dione, 3-tert-butyldimethylsilyloxy glutaricanhydride, 3-tert-butyldimethylsiloxy pentanedioic anhydride, 3-tert-butyldimethyl silyl oxy glutaric anhydride
4-[tert-butyl(dimethyl)silyl]oxyoxane-2,6-dione
Safety and Handling

Safety and Handling

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses, if present and easy to do.
Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Warning

storageNote1 : 2° to 8°C

Documents