missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

3-tert-Butyl-4-hydroxyanisole, 98%, ACROS Organics™

Manufacturer:  Thermo Fisher Scientific AC439691000

 View more versions of this product

Catalog No. AC439691000



Specifications

Specifications

3-tert-Butyl-4-hydroxyanisole
Authentic
White
100g
97.5
97.5% min. (GC)
2-1,1-dimethylethyl-4-methoxyphenol, 2-butyl-4-hydroxyanisole, 2-tert-butyl-4-methoxyphenol, 3-bha, 3-t-butyl-4-hydroxyanisole, 3-tert-butyl-4-hydroxyanisole, 4-hydroxy-3-tert-butylanisole, 4-methoxy-2-tert-butylphenol, phenol, 2-1,1-dimethylethyl-4-methoxy, phenol, 2-tert-butyl-4-methoxy
MRBKEAMVRSLQPH-UHFFFAOYSA-N
2-tert-butyl-4-methoxyphenol
8456
180.25
98%
110°C
Glass bottle
58.0°C to 66.0°C
121-00-6
100.0
C11H16O2
Solubility in water: in water:(20°C) insoluble.,in ethanol:(20°C) soluble
CC(C)(C)C1=C(C=CC(=C1)OC)O
180.25
CHEBI:76358
Powder/Waxy Solid

Chemical Identifiers

121-00-6
180.25
2-1,1-dimethylethyl-4-methoxyphenol, 2-butyl-4-hydroxyanisole, 2-tert-butyl-4-methoxyphenol, 3-bha, 3-t-butyl-4-hydroxyanisole, 3-tert-butyl-4-hydroxyanisole, 4-hydroxy-3-tert-butylanisole, 4-methoxy-2-tert-butylphenol, phenol, 2-1,1-dimethylethyl-4-methoxy, phenol, 2-tert-butyl-4-methoxy
CHEBI:76358
CC(C)(C)C1=C(C=CC(=C1)OC)O
C11H16O2
MRBKEAMVRSLQPH-UHFFFAOYSA-N
8456
2-tert-butyl-4-methoxyphenol
Safety and Handling

Safety and Handling

GHS H Statement
Causes skin irritation.
May cause an allergic skin reaction.
Causes serious eye irritation.

GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN: Wash with plenty of soap and water.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses,if present a

GHS Signal Word: Warning

EINECSNumber : 204-442-7

SDS Documents