Please login to your online account to display your discounted pricing

Alfa Aesar™ 3-tert-Butyl-2-pyrazolin-5-one, 98%

$47.32 - $150.68

Chemical Identifiers

Chemical Name or Material 3-tert-Butyl-2-pyrazolin-5-one
CAS 29211-68-5
Molecular Formula C7H12N2O
Formula Weight 140.19
MDL Number MFCD00051760
Beilstein 775174
Synonym 5-tert-Butyl-2,4-dihydro-3H-pyrazol-3-one
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAL0776103

  SDS
alfa aesar™
L0776103
1g Each for $47.32

AAL0776106

  SDS
alfa aesar™
L0776106
5g Each for $150.68
Description & Specifications

Specifications

Chemical Name or Material 3-tert-Butyl-2-pyrazolin-5-one
CAS 29211-68-5
Assay 98%
Molecular Formula C7H12N2O
Formula Weight 140.19
MDL Number MFCD00051760
Beilstein 775174
Synonym 5-tert-Butyl-2,4-dihydro-3H-pyrazol-3-one
Melting Point 208° to 212°C
Quantity 1g