Please login to your online account to display your discounted pricing

Alfa Aesar™ 3-tert-Butyl-1-phenyl-2-pyrazolin-5-one, 98%

$36.32 - $36.32

Chemical Identifiers

Chemical Name or Material 3-tert-Butyl-1-phenyl-2-pyrazolin-5-one
CAS 6631-89-6
Molecular Formula C13H16N2O
Formula Weight 216.29
MDL Number MFCD00051762
Beilstein 4804384
Synonym 5-tert-Butyl-2,4-dihydro-3H-pyrazol-3-one
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAB2193003

 
alfa aesar™
B2193003
1g Each for $36.32
Description & Specifications

Specifications

Formula Weight 216.29
Chemical Name or Material 3-tert-Butyl-1-phenyl-2-pyrazolin-5-one
Assay 98%
Molecular Formula C13H16N2O
MDL Number MFCD00051762
CAS 6631-89-6
Beilstein 4804384
Synonym 5-tert-Butyl-2,4-dihydro-3H-pyrazol-3-one
Melting Point 108° to 113°C
Quantity 1g