missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

3-(tert-Butoxycarbonylamino)-1-propanol, TCI America™

$47.36 - $142.09

Chemical Identifiers

CAS 58885-58-8
Molecular Formula C8H17NO3
Molecular Weight (g/mol) 175.228
MDL Number MFCD00191883
InChI Key XDJCYKMWJCYQJM-UHFFFAOYSA-N
Synonym 3-boc-amino-1-propanol, tert-butyl n-3-hydroxypropyl carbamate, boc-beta-ala-ol, 3-tert-butoxycarbonylamino-1-propanol, tert-butyl 3-hydroxypropylcarbamate, 3-n-boc-amino propanol, tert-butyl 3-hydroxypropyl carbamate, n-3-hydroxypropyl carbamic acid tert-butyl ester, tert-butyl 3-hydroxypropyl carbamate, 3-hydroxy-propyl-carbamic acid tert-butyl ester
PubChem CID 3838134
IUPAC Name tert-butyl N-(3-hydroxypropyl)carbamate
SMILES CC(C)(C)OC(=O)NCCCO
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
H09005ML View Documents TCI America
H09005ML
5mL Each for $47.36
 
H090025ML View Documents TCI America
H090025ML
25mL Each for $142.09
 
Specifications

Chemical Identifiers

58885-58-8
175.228
XDJCYKMWJCYQJM-UHFFFAOYSA-N
3838134
CC(C)(C)OC(=O)NCCCO
C8H17NO3
MFCD00191883
3-boc-amino-1-propanol, tert-butyl n-3-hydroxypropyl carbamate, boc-beta-ala-ol, 3-tert-butoxycarbonylamino-1-propanol, tert-butyl 3-hydroxypropylcarbamate, 3-n-boc-amino propanol, tert-butyl 3-hydroxypropyl carbamate, n-3-hydroxypropyl carbamic acid tert-butyl ester, tert-butyl 3-hydroxypropyl carbamate, 3-hydroxy-propyl-carbamic acid tert-butyl ester
tert-butyl N-(3-hydroxypropyl)carbamate

Specifications

Colorless
58885-58-8
MFCD00191883
XDJCYKMWJCYQJM-UHFFFAOYSA-N
tert-butyl N-(3-hydroxypropyl)carbamate
3838134
Clear Liquid at 20°C
3-(tert-Butoxycarbonylamino)-1-propanol
5mL
C8H17NO3
3-boc-amino-1-propanol, tert-butyl n-3-hydroxypropyl carbamate, boc-beta-ala-ol, 3-tert-butoxycarbonylamino-1-propanol, tert-butyl 3-hydroxypropylcarbamate, 3-n-boc-amino propanol, tert-butyl 3-hydroxypropyl carbamate, n-3-hydroxypropyl carbamic acid tert-butyl ester, tert-butyl 3-hydroxypropyl carbamate, 3-hydroxy-propyl-carbamic acid tert-butyl ester
CC(C)(C)OC(=O)NCCCO
175.228
175.23
≥96.0% (GC)
Safety and Handling

Safety and Handling

TSCA : No