Please login to your online account to display your discounted pricing

Alfa Aesar™ 3-Sulfobenzoic acid disodium salt monohydrate, 97%

$36.61 - $236.57

Chemical Identifiers

Chemical Name or Material 3-Sulfobenzoic acid disodium salt monohydrate
CAS 14995-40-5
Molecular Formula C7H4Na2O5S·H2O
Formula Weight 264.17 (246.15 Anhydrous)
MDL Number MFCD00149379
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAB2415414

  SDS
alfa aesar™
B2415414
25g Each for $36.61

AAB2415422

  SDS
alfa aesar™
B2415422
100g Each for $74.89

AAB2415436

  SDS
alfa aesar™
B2415436
500g Each for $236.57
Description & Specifications

Specifications

Formula Weight 264.17 (246.15 Anhydrous)
Quantity 25g
Chemical Name or Material 3-Sulfobenzoic acid disodium salt monohydrate
MDL Number MFCD00149379
Molecular Formula C7H4Na2O5S·H2O
Assay 97%
CAS 14995-40-5