missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ 3-Sulfo-N-succinimidyl 4-(maleimidomethyl)cyclohexane-1-carboxylate sodium salt, 97+%

Manufacturer:  Alfa Aesar™ H54807MC

 View more versions of this product

Catalog No. AAH54807MC



Description

Description

Applications
3-Sulfo-N-succinimidyl 4-(maleimidomethyl)cyclohexane-1-carboxylate sodium salt is a cross-linking reagent incorporating an extended spacer with amine and sulfhydryl reactivity. Typically, coupled initially to molecules containing primary amines by amide bonds buffered at pH 7.5 (6.5-8.5). Second coupling specific for molecules containing free sulfhydryl by thioether linkage buffered at pH 6.8 (6.5-7.0). Useful for preparation of enzyme immunoconjugates and hapten carrier molecule conjugates.

Solubility
Soluble in water (2.5 mg/ml) and dimethyl formamide.

Notes
Keep container tightly sealed. Store in cool, dry conditions in well sealed containers. Incompatible with oxidizing agents.
Specifications

Specifications

3-Sulfo-N-succinimidyl 4-(maleimidomethyl)cyclohexane-1-carboxylate sodium salt
92921-24-9
MFCD00054978
Soluble in water (2.5mg/ml) and dimethyl formamide.
C1CC(CCC1CN2C(=O)C=CC2=O)C(=O)ON3C(=O)CC(C3=O)S(=O)(=O)[O-].[Na+]
436.367
436.37
100mg
C16H17N2NaO9S
3-sulfo-n-succinimidyl 4-n-maleimidomethyl cyclohexane-1-carboxylate sodium salt, 4-maleimidomethyl cyclohexane-1-carboxylic acid 3-sulfo-n-hydroxysuccinimide ester sodium salt, bicl207, sodium 1-4-2,5-dioxo-2,5-dihydro-1h-pyrrol-1-yl methyl cyclohexanecarbonyl oxy-2,5-dioxopyrrolidine-3-sulfonate, sulfo-n-succinimidyl 4-maleimidomethyl cyclohexane-1-carboxylate sodium salt, sulfo-smcc, sulfo-smcc crosslinker, sulfo-smcc sodium, sulfo-smcc sodium salt, sulfo-trans-smcc
VUFNRPJNRFOTGK-UHFFFAOYSA-M
sodium;1-[4-[(2,5-dioxopyrrol-1-yl)methyl]cyclohexanecarbonyl]oxy-2,5-dioxopyrrolidine-3-sulfonate
4618794
≥98%

Chemical Identifiers

92921-24-9
436.367
VUFNRPJNRFOTGK-UHFFFAOYSA-M
4618794
C1CC(CCC1CN2C(=O)C=CC2=O)C(=O)ON3C(=O)CC(C3=O)S(=O)(=O)[O-].[Na+]
C16H17N2NaO9S
MFCD00054978
3-sulfo-n-succinimidyl 4-n-maleimidomethyl cyclohexane-1-carboxylate sodium salt, 4-maleimidomethyl cyclohexane-1-carboxylic acid 3-sulfo-n-hydroxysuccinimide ester sodium salt, bicl207, sodium 1-4-2,5-dioxo-2,5-dihydro-1h-pyrrol-1-yl methyl cyclohexanecarbonyl oxy-2,5-dioxopyrrolidine-3-sulfonate, sulfo-n-succinimidyl 4-maleimidomethyl cyclohexane-1-carboxylate sodium salt, sulfo-smcc, sulfo-smcc crosslinker, sulfo-smcc sodium, sulfo-smcc sodium salt, sulfo-trans-smcc
sodium;1-[4-[(2,5-dioxopyrrol-1-yl)methyl]cyclohexanecarbonyl]oxy-2,5-dioxopyrrolidine-3-sulfonate
Safety and Handling

Safety and Handling

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c

H315-H319-H335

TSCA : No

Recommended Storage : Keep cold

SDS
Documents