missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

3-Quinuclidinone Hydrochloride 98.0+%, TCI America™

Manufacturer:  Tci America Q005225G

 View more versions of this product

Catalog No. Q005225G



Specifications

Specifications

3-Quinuclidinone Hydrochloride
300°C
1193-65-3
MFCD00137391
RFDPHKHXPMDJJD-UHFFFAOYSA-N
1-azabicyclo[2.2.2]octan-3-one;hydrochloride
102019
Crystal-Powder at 20°C
White-Yellow
25g
C7H12ClNO
1-aza-3-oxobicyclo 2.2.2 octane
C1CN2CCC1C(=O)C2.Cl
161.629
161.63
≥98.0% (T)

Chemical Identifiers

1193-65-3
161.629
RFDPHKHXPMDJJD-UHFFFAOYSA-N
102019
C1CN2CCC1C(=O)C2.Cl
C7H12ClNO
MFCD00137391
1-aza-3-oxobicyclo 2.2.2 octane
1-azabicyclo[2.2.2]octan-3-one;hydrochloride
Safety and Handling

Safety and Handling

TSCA : Yes

Documents