Please login to your online account to display your discounted pricing

3-Pyrroline, ACROS Organics™


Quantity 250mg
Packaging Glass Ampule with Screw-cap

Manufacturer: thermo fisher scientific  AC132132500

 View more versions of this product

Catalog No. AC132132500

 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description & Specifications

Chemical Identifiers

Physical Form Liquid
Assay ≥96.0%
Formula Weight 69.11
Quantity 250mg
Color Clear colorless to light yellow
Packaging Glass Ampule with Screw-cap
Density 0.9100g/mL
Merck Index 15, 8131
CAS Min % 96.0
CAS Max % 100.0
Beilstein 20, V,4, 274
Infrared Spectrum Authentic
Refractive Index 1.4690 to 1.4710 (20°C, 589nm)
Assay Percent Range 97%
MDL Number MFCD00005213
Synonym 2, 5-Dihydro-1H-pyrrole
Health Hazard 3 GHS P Statement:
IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
Wear eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Immediately call a POISON CENTER or doctor/physician.
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
Ground/Bond container and receiving equipment.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
Chemical Name or Material 3-Pyrroline
Molecular Formula C4H7N
CAS 109-96-6
Health Hazard 1 GHS Signal Word: Danger
Health Hazard 2 GHS H Statement:
Causes severe skin burns and eye damage.
Causes serious eye damage.
Highly flammable liquid and vapor.

Specifications

Chemical Name or Material 3-Pyrroline
CAS 109-96-6
CAS Min % 96.0
CAS Max % 100.0
Assay ≥96.0%
Assay Percent Range 97%
Molecular Formula C4H7N
Formula Weight 69.11
MDL Number MFCD00005213
Synonym 2, 5-Dihydro-1H-pyrrole
Accessories

GHS Signal Word: Danger

GHS P Statement:
IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
Wear eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Immediately call a POISON CENTER or doctor/physician.
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
Ground/Bond container and receiving equipment.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS H Statement:
Causes severe skin burns and eye damage.
Causes serious eye damage.
Highly flammable liquid and vapor.

SDS & Safety Documents