missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

3-Pyrroline, ACROS Organics™

Chemical Identifiers

CAS 109-96-6
CAS Min % 96.0
CAS Max % 100.0
Assay Percent Range 97%
Molecular Formula C4H7N
Formula Weight 69.11
MDL Number MFCD00005213
Synonym 2, 5-Dihydro-1H-pyrrole
Chemical Name or Material 3-Pyrroline
Assay ≥96.0%
View More Specs


Products ${productFamilyLength}
Catalog Number Mfr. No. Packaging Quantity Price Quantity  
Catalog Number Mfr. No. Packaging Quantity Price Quantity  
AC132132500 SDS Thermo Fisher Scientific
AC132132500
Glass Ampule with Screw-cap 250mg N/A N/A  
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Specifications Description & Specifications

Chemical Identifiers

109-96-6
100.0
C4H7N
MFCD00005213
3-Pyrroline
96.0
97%
69.11
2, 5-Dihydro-1H-pyrrole
≥96.0%

Specifications

69.11
250mg
Glass Ampule with Screw-cap
15, 8131
96.0
97%
1.4690 to 1.4710 (20°C, 589nm)
Authentic
C4H7N
≥96.0%
Liquid
Clear colorless to light yellow
0.9100g/mL
100.0
20, V,4, 274
MFCD00005213
2, 5-Dihydro-1H-pyrrole
109-96-6
3-Pyrroline

Safety and Handling

GHS H Statement:
Causes severe skin burns and eye damage.
Causes serious eye damage.
Highly flammable liquid and vapor.

GHS P Statement:
IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
Wear eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Immediately call a POISON CENTER or doctor/physician.
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
Ground/Bond container and receiving equipment.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS Signal Word: Danger