Please login to your online account to display your discounted pricing

3-(Pyrrolidin-1-ylmethyl)benzoic acid hydrochloride, 90%, Maybridge


Quantity 5g
Packaging Amber Glass Bottle

Manufacturer: fisher scientific company  CC55751DE

 View more versions of this product

Catalog No. CC55751DE

 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description & Specifications

Chemical Identifiers

Formula Weight 241.72
Quantity 5g
Assay 90%
Chemical Name or Material 3-(Pyrrolidin-1-ylmethyl)benzoic acid hydrochloride
Packaging Amber Glass Bottle
Assay Percent Range 90%
Molecular Formula C12H15NO2·ClH
CAS 887922-93-2
Health Hazard 1 Warning

Specifications

Chemical Name or Material 3-(Pyrrolidin-1-ylmethyl)benzoic acid hydrochloride
Assay 90%
Assay Percent Range 90%
Molecular Formula C12H15NO2·ClH
Formula Weight 241.72
Accessories

Warning

SDS & Safety Documents