Please login to your online account to display your discounted pricing

3-(Pyrrolidin-1-ylmethyl)aniline, 97%, Maybridge

Chemical Identifiers

Chemical Name or Material 3-(Pyrrolidin-1-ylmethyl)aniline
Assay 97%
Assay Percent Range 97%
Molecular Formula C11H16N2
Formula Weight 176.26
View More Specs
Products
Catalog Number Mfr. No. Packaging Quantity Price Quantity    

CC55714DE

 
fisher scientific company
CC55714DE
Amber Glass Bottle 5g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CC55714DA

  SDS
fisher scientific company
CC55714DA
Amber Glass Bottle 1g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CC55714CB

  SDS
fisher scientific company
CC55714CB
CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Formula Weight 176.26
Quantity 5g
Assay 97%
Chemical Name or Material 3-(Pyrrolidin-1-ylmethyl)aniline
Packaging Amber Glass Bottle
Assay Percent Range 97%
Molecular Formula C11H16N2
CAS 183365-31-3