missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
3-(Pyrrolidin-1-ylmethyl)aniline, 97%, Thermo Scientific™
Supplier: Thermo Scientific Chemicals CC55714DE
Specifications
3-(Pyrrolidin-1-ylmethyl)aniline | |
97% | |
C11H16N2 | |
3-pyrrolidin-1-ylmethyl aniline, 3-pyrrolidin-1-ylmethyl-phenylamine, benzenamine, 3-1-pyrrolidinylmethyl, 3-1-pyrrolidinylmethyl aniline, 3-pyrrolidinylmethyl phenylamine, 3-pyrrolidinylmethylaniline, 3-pyrrolidin-1-ylmethyl-aniline, 3-1-pyrrolidinylmethyl benzenamine, 3-pyrrolidin-1-yl-methyl-aniline, 3-pyrrolidin-1-yl methyl aniline | |
C1CCN(C1)CC2=CC(=CC=C2)N | |
176.263 | |
176.26 |
183365-31-3 | |
Amber Glass Bottle | |
5g | |
RZDBCURLSQNEAD-UHFFFAOYSA-N | |
3-(pyrrolidin-1-ylmethyl)aniline | |
1096407 | |
97% |
Chemical Identifiers
183365-31-3 | |
176.263 | |
3-pyrrolidin-1-ylmethyl aniline, 3-pyrrolidin-1-ylmethyl-phenylamine, benzenamine, 3-1-pyrrolidinylmethyl, 3-1-pyrrolidinylmethyl aniline, 3-pyrrolidinylmethyl phenylamine, 3-pyrrolidinylmethylaniline, 3-pyrrolidin-1-ylmethyl-aniline, 3-1-pyrrolidinylmethyl benzenamine, 3-pyrrolidin-1-yl-methyl-aniline, 3-pyrrolidin-1-yl methyl aniline | |
3-(pyrrolidin-1-ylmethyl)aniline |
C11H16N2 | |
RZDBCURLSQNEAD-UHFFFAOYSA-N | |
1096407 | |
C1CCN(C1)CC2=CC(=CC=C2)N |