Please login to your online account to display your discounted pricing

3-Pyrrolidin-1-ylaniline, 97%, Maybridge


Quantity 10g
Packaging Amber Glass Bottle

Manufacturer: fisher scientific company  CC38514EA

 View more versions of this product

Catalog No. CC38514EA

 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description & Specifications

Chemical Identifiers

Formula Weight 162.23
Quantity 10g
Assay 97%
CAS 115833-93-7
Chemical Name or Material 3-Pyrrolidin-1-ylaniline
Packaging Amber Glass Bottle
Assay Percent Range 97%
Molecular Formula C10H14N2

Specifications

Chemical Name or Material 3-Pyrrolidin-1-ylaniline
Assay 97%
Assay Percent Range 97%
Molecular Formula C10H14N2
Formula Weight 162.23