Learn More
3-Pyrimidin-2-ylbenzylamine, 97%, Thermo Scientific™
Chemical Identifiers
CAS | 910036-92-9 |
---|---|
Molecular Formula | C11H11N3 |
Molecular Weight (g/mol) | 185.23 |
MDL Number | MFCD09702384 |
InChI Key | MXSXDKVANPZCBQ-UHFFFAOYSA-N |
Synonym | 3-pyrimidin-2-ylbenzylamine, 3-pyrimidin-2-yl phenyl methanamine, 3-pyrimidin-2-yl benzylamine, 1-3-pyrimidin-2-yl phenyl methanamine, 3-pyrimidin-2-ylphenyl methylamine, benzenemethanamine, 3-2-pyrimidinyl |
PubChem CID | 24229566 |
IUPAC Name | (3-pyrimidin-2-ylphenyl)methanamine |
SMILES | NCC1=CC=CC(=C1)C1=NC=CC=N1 |
Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | ||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | ||||
CC56313CB
|
Thermo Scientific Chemicals
CC56313CB |
250mg | Amber Glass Bottle |
N/A
|
N/A | ||||
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance. | |||||||||
CC56313DA
|
Thermo Scientific Chemicals
CC56313DA |
1g | Amber Glass Bottle |
N/A
|
N/A | ||||
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance. | |||||||||
Chemical Identifiers
910036-92-9 | |
185.23 | |
MXSXDKVANPZCBQ-UHFFFAOYSA-N | |
24229566 | |
NCC1=CC=CC(=C1)C1=NC=CC=N1 |
C11H11N3 | |
MFCD09702384 | |
3-pyrimidin-2-ylbenzylamine, 3-pyrimidin-2-yl phenyl methanamine, 3-pyrimidin-2-yl benzylamine, 1-3-pyrimidin-2-yl phenyl methanamine, 3-pyrimidin-2-ylphenyl methylamine, benzenemethanamine, 3-2-pyrimidinyl | |
(3-pyrimidin-2-ylphenyl)methanamine |
Specifications
910036-92-9 | |
100.0 | |
Amber Glass Bottle | |
MFCD09702384 | |
3-pyrimidin-2-ylbenzylamine, 3-pyrimidin-2-yl phenyl methanamine, 3-pyrimidin-2-yl benzylamine, 1-3-pyrimidin-2-yl phenyl methanamine, 3-pyrimidin-2-ylphenyl methylamine, benzenemethanamine, 3-2-pyrimidinyl | |
NCC1=CC=CC(=C1)C1=NC=CC=N1 | |
185.23 | |
185.23 | |
3-Pyrimidin-2-ylbenzylamine |
94.5 | |
95% | |
C11H11N3 | |
250mg | |
MXSXDKVANPZCBQ-UHFFFAOYSA-N | |
(3-pyrimidin-2-ylphenyl)methanamine | |
24229566 | |
95% |
Safety and Handling
GHS H Statement:
Harmful if swallowed.
Harmful in contact with skin.
Causes severe skin burns and eye damage.
Causes serious eye damage.
GHS P Statement:
Wear protective gloves/protective clothing/eye protection/face protection.
IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
Wear eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Immediately call a POISON CENTER or doctor/physician.
IF ON SKIN: Wash with plenty of soap and water.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
GHS Signal Word: Danger