Learn More
3-Pyridylacetonitrile, 98%, Thermo Scientific™
Chemical Identifiers
CAS | 6443-85-2 |
---|---|
Molecular Formula | C7H6N2 |
Molecular Weight (g/mol) | 118.14 |
MDL Number | MFCD00006406 |
InChI Key | OIPHWUPMXHQWLR-UHFFFAOYSA-N |
Synonym | 2-3-pyridyl ethanenitrile, 2-pyridin-3-yl acetonitrile, 3-cyanomethyl pyridine, 3-cyanomethylpyridine, 3-pyridineacetonitrile, 3-pyridyl acetonitrile, 3-pyridylacetonitrile, pyridin-3-yl-acetonitrile, pyridin-3-ylacetonitrile, pyridine-3-acetonitrile |
PubChem CID | 80923 |
IUPAC Name | 2-pyridin-3-ylacetonitrile |
SMILES | N#CCC1=CC=CN=C1 |
Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | ||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | ||||
AC131970010
|
Thermo Fisher Scientific
AC131970010 |
1g | Glass bottle |
N/A
|
N/A | ||||
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance. | |||||||||
AC131970050
|
Thermo Fisher Scientific
AC131970050 |
5g | Glass bottle |
N/A
|
N/A | ||||
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance. | |||||||||
AC131970250
|
Thermo Fisher Scientific
AC131970250 |
25g | Glass bottle |
N/A
|
N/A | ||||
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance. | |||||||||
Description
This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
Chemical Identifiers
6443-85-2 | |
118.14 | |
OIPHWUPMXHQWLR-UHFFFAOYSA-N | |
80923 | |
N#CCC1=CC=CN=C1 |
C7H6N2 | |
MFCD00006406 | |
2-3-pyridyl ethanenitrile, 2-pyridin-3-yl acetonitrile, 3-cyanomethyl pyridine, 3-cyanomethylpyridine, 3-pyridineacetonitrile, 3-pyridyl acetonitrile, 3-pyridylacetonitrile, pyridin-3-yl-acetonitrile, pyridin-3-ylacetonitrile, pyridine-3-acetonitrile | |
2-pyridin-3-ylacetonitrile |
Specifications
6443-85-2 | |
100.0 | |
Colorless to Yellow | |
98°C | |
Authentic | |
(C5H4N)CH2C∽N | |
Glass bottle | |
2-3-pyridyl ethanenitrile, 2-pyridin-3-yl acetonitrile, 3-cyanomethyl pyridine, 3-cyanomethylpyridine, 3-pyridineacetonitrile, 3-pyridyl acetonitrile, 3-pyridylacetonitrile, pyridin-3-yl-acetonitrile, pyridin-3-ylacetonitrile, pyridine-3-acetonitrile | |
OIPHWUPMXHQWLR-UHFFFAOYSA-N | |
N#CCC1=CC=CN=C1 | |
118.14 | |
118.14 | |
98% |
97.5 | |
1.1000g/mL | |
105°C to 108°C (1.5mmHg) | |
97.5 % min. (GC) | |
C7H6N2 | |
MFCD00006406 | |
1.5289 | |
1g | |
1.1 | |
2-pyridin-3-ylacetonitrile | |
80923 | |
Liquid | |
3-Pyridylacetonitrile |
Safety and Handling
GHS H Statement
Harmful in contact with skin.
Harmful if inhaled.
May cause respiratory irritation.
Harmful if swallowed.
Causes skin irritation.
Causes serious eye irritation.
GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN: Wash with plenty of soap and water.
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Call a POISON CENTER or doctor
GHS Signal Word: Warning
EINECSNumber : 229-241-1
RUO – Research Use Only