missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

3-Pyridylacetonitrile, 98%, ACROS Organics™

Manufacturer:  Thermo Fisher Scientific AC131970050

 View more versions of this product

Catalog No. AC131970050

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description

Description

Specifications

Specifications

3-Pyridylacetonitrile
98°C
Glass bottle
1.1
Colorless to Yellow
6443-85-2
100.0
C7H6N2
2-3-pyridyl ethanenitrile, 2-pyridin-3-yl acetonitrile, 3-cyanomethyl pyridine, 3-cyanomethylpyridine, 3-pyridineacetonitrile, 3-pyridyl acetonitrile, 3-pyridylacetonitrile, pyridin-3-yl-acetonitrile, pyridin-3-ylacetonitrile, pyridine-3-acetonitrile
C1=CC(=CN=C1)CC#N
118.14
118.14
98%
1.1000g/mL
Authentic
1.5289
105°C to 108°C (1.5mmHg)
5g
97.5
97.5 % min. (GC)
(C5H4N)CH2C∽N
OIPHWUPMXHQWLR-UHFFFAOYSA-N
2-pyridin-3-ylacetonitrile
80923
Liquid

Chemical Identifiers

6443-85-2
118.14
2-3-pyridyl ethanenitrile, 2-pyridin-3-yl acetonitrile, 3-cyanomethyl pyridine, 3-cyanomethylpyridine, 3-pyridineacetonitrile, 3-pyridyl acetonitrile, 3-pyridylacetonitrile, pyridin-3-yl-acetonitrile, pyridin-3-ylacetonitrile, pyridine-3-acetonitrile
2-pyridin-3-ylacetonitrile
C7H6N2
OIPHWUPMXHQWLR-UHFFFAOYSA-N
80923
C1=CC(=CN=C1)CC#N
Safety and Handling

Safety and Handling

GHS H Statement
Harmful in contact with skin.
Harmful if inhaled.
May cause respiratory irritation.
Harmful if swallowed.
Causes skin irritation.
Causes serious eye irritation.

GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN: Wash with plenty of soap and water.
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Call a POISON CENTER or doctor

GHS Signal Word: Warning

EINECSNumber : 229-241-1

SDS
Documents