Learn More
3-Pyridylacetonitrile, 98%, Thermo Scientific™
Supplier: Thermo Scientific Chemicals 131970010
Description
This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
Specifications
3-Pyridylacetonitrile | |
97.5 | |
1.1000g/mL | |
98°C | |
97.5 % min. (GC) | |
C7H6N2 | |
(C5H4N)CH2C∽N | |
1g | |
3-pyridineacetonitrile, 3-pyridylacetonitrile, 2-pyridin-3-yl acetonitrile, pyridine-3-acetonitrile, 3-cyanomethylpyridine, 3-cyanomethyl pyridine, pyridin-3-ylacetonitrile, 2-3-pyridyl ethanenitrile, pyridin-3-yl-acetonitrile, 3-pyridyl acetonitrile | |
N#CCC1=CC=CN=C1 | |
118.14 | |
118.14 |
6443-85-2 | |
100.0 | |
105°C to 108°C (1.5mmHg) | |
Authentic | |
Glass bottle | |
1.5289 | |
MFCD00006406 | |
1.1 | |
OIPHWUPMXHQWLR-UHFFFAOYSA-N | |
2-pyridin-3-ylacetonitrile | |
80923 | |
98% |
Chemical Identifiers
6443-85-2 | |
118.14 | |
OIPHWUPMXHQWLR-UHFFFAOYSA-N | |
80923 | |
N#CCC1=CC=CN=C1 |
C7H6N2 | |
MFCD00006406 | |
3-pyridineacetonitrile, 3-pyridylacetonitrile, 2-pyridin-3-yl acetonitrile, pyridine-3-acetonitrile, 3-cyanomethylpyridine, 3-cyanomethyl pyridine, pyridin-3-ylacetonitrile, 2-3-pyridyl ethanenitrile, pyridin-3-yl-acetonitrile, 3-pyridyl acetonitrile | |
2-pyridin-3-ylacetonitrile |
Safety and Handling
GHS H Statement
Harmful in contact with skin.
Harmful if inhaled.
May cause respiratory irritation.
Harmful if swallowed.
Causes skin irritation.
Causes serious eye irritation.
GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN: Wash with plenty of soap and water.
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Call a POISON CENTER or doctor
GHS Signal Word: Warning
EINECSNumber : 229-241-1
RUO â Research Use Only