Please login to your online account to display your discounted pricing

3-Pyridylacetic acid hydrochloride, 98%, ACROS Organics™


Quantity 2.5g
Packaging Glass bottle

Manufacturer: fisher scientific acros  131960025

 View more versions of this product

Catalog No. AC131960025

  • / Each
 

Description & Specifications

Chemical Identifiers

Assay ≥97.5%
Formula Weight 173.6
Physical Form Crystalline Powder, Crystals and/or Chunks
Quantity 2.5g
Color White to light yellow or beige
Merck Index 10, 4635
CAS Max % 100.0
Linear Formula (C5H4N)CH2OOH·HCl
Health Hazard 2 GHS H Statement:
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Health Hazard 3 GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
Chemical Name or Material 3-Pyridylacetic acid hydrochloride
Molecular Formula C7H7NO2·ClH
MDL Number MFCD00012819
CAS Min % 97.5
Assay Percent Range 98%
CAS 6419-36-9
Packaging Glass bottle
Synonym 3-Pyridineacetic acid
Infrared Spectrum Authentic

Specifications

Chemical Name or Material 3-Pyridylacetic acid hydrochloride
CAS 6419-36-9
CAS Min % 97.5
CAS Max % 100.0
Assay ≥97.5%
Assay Percent Range 98%
Molecular Formula C7H7NO2·ClH
Linear Formula (C5H4N)CH2OOH·HCl
Formula Weight 173.6
MDL Number MFCD00012819
Synonym 3-Pyridineacetic acid
Accessories

GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS H Statement:
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

SDS & Safety Documents