missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

3-Pyridyl isothiocyanate, ≥95%, Thermo Scientific™

Supplier:  Thermo Scientific Chemicals CC04107DA

 View more versions of this product

Catalog No. CC04107DA



Description

Description

Specifications

Specifications

3-Pyridyl isothiocyanate
94.5
1.22g/mL
Amber Glass Bottle
MFCD00052351
3-pyridyl isothiocyanate, pyridine, 3-isothiocyanato, 3-isothiocyanato-pyridine, pyridine-3-isothiocyanate, pyridine,3-isothiocyanato, 3-pyridylisothiocyanate, 3-pyridyl isotiocyanate, acmc-20an1z, 3-pyridyl-isothiocyanate, pyridin-3-isothiocyanate
C1=CC(=CN=C1)N=C=S
136.172
136.18
17452-27-6
100.0
95%
C6H4N2S
1 g
VMSZFBSYWXMXRF-UHFFFAOYSA-N
3-isothiocyanatopyridine
2737242
95%

Chemical Identifiers

17452-27-6
136.172
VMSZFBSYWXMXRF-UHFFFAOYSA-N
2737242
C1=CC(=CN=C1)N=C=S
C6H4N2S
MFCD00052351
3-pyridyl isothiocyanate, pyridine, 3-isothiocyanato, 3-isothiocyanato-pyridine, pyridine-3-isothiocyanate, pyridine,3-isothiocyanato, 3-pyridylisothiocyanate, 3-pyridyl isotiocyanate, acmc-20an1z, 3-pyridyl-isothiocyanate, pyridin-3-isothiocyanate
3-isothiocyanatopyridine
Safety and Handling

Safety and Handling

GHS H Statement:
Harmful if swallowed.
Causes serious eye irritation.
May cause respiratory irritation.
Harmful if inhaled.
Harmful in contact with skin.
Causes skin irritation.

GHS P Statement:
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN: Wash with plenty of soap and water.
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS Signal Word: Warning

SDS Documents

Documents

Promotions

Promotions