Learn More
3-Pyridinecarboxaldehyde, 98%, Thermo Scientific™
Supplier: Thermo Scientific Chemicals 131835000
Description
This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
Specifications
3-Pyridinecarboxaldehyde | |
1.1420g/mL | |
84°C | |
97.5% min. (GC) | |
C6H5NO | |
(C5H4N)COH | |
21,I,288 | |
3-pyridinecarboxaldehyde, nicotinaldehyde, 3-pyridinaldehyde, pyridine-3-carboxaldehyde, 3-pyridylcarboxaldehyde, 3-pyridylaldehyde, nicotinealdehyde, 3-formylpyridine, pyridine-3-aldehyde, nicotinic aldehyde | |
QJZUKDFHGGYHMC-UHFFFAOYSA-N | |
pyridine-3-carbaldehyde | |
10371 | |
107.11 |
500-22-1 | |
95.0°C to 97.0°C (15.0mmHg) | |
Authentic | |
Glass bottle | |
1.5480 to 1.5500 | |
500g | |
1.142 | |
Solubility in water: miscible. Other solubilities: soluble in most organic solvents | |
C1=CC(=CN=C1)C=O | |
107.11 | |
CHEBI:28345 | |
98% |
Safety and Handling
GHS H Statement
May cause an allergic skin reaction.
Causes severe skin burns and eye damage.
May cause allergy or asthma symptoms or breathing difficulties if inhaled.
GHS P Statement
IF SWALLOWED: rinse mouth.
Do NOT induce vomiting.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses,if present and easy to do.
Continue rinsing.
Immediately call a POISON CENTER
GHS Signal Word: Danger
EINECSNumber : 207-900-4
RUO â Research Use Only