Please login to your online account to display your discounted pricing

3-Pyridinealdoxime, 98%, ACROS Organics™

Chemical Identifiers

Chemical Name or Material 3-Pyridinealdoxime
Name Note 98%
CAS 1193-92-6
CAS Min % 97.5
CAS Max % 100.0
Assay ≥97.5%
Assay Percent Range 98%
Molecular Formula C6H6N2O
Formula Weight 122.13
MDL Number MFCD00006409
View More Specs
Products
Catalog Number Mfr. No. Packaging Quantity Price Quantity    

AC131800050

  SDS
thermo fisher scientific
131800050
Glass bottle 5g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Physical Form Fine Crystalline Powder
Assay ≥97.5%
Formula Weight 122.13
Quantity 5g
Color White
RTECS Number UT4060000
Melting Point 149° to 153°C
EINECS Number 214-782-8
TSCA TSCA
CAS Max % 100.0
Health Hazard 2 GHS H Statement:
Harmful if swallowed.
May cause respiratory irritation.
Causes serious eye irritation.
Causes skin irritation.
Harmful if inhaled.
Health Hazard 3 GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
Chemical Name or Material 3-Pyridinealdoxime
Packaging Glass bottle
Name Note 98%
Assay Percent Range 98%
Molecular Formula C6H6N2O
MDL Number MFCD00006409
CAS Min % 97.5
Infrared Spectrum Authentic
CAS 1193-92-6