Please login to your online account to display your discounted pricing

Alfa Aesar™ 3-Pyridineacetic acid hydrochloride, 98+%

$114.66 - $1,090.89

Chemical Identifiers

Chemical Name or Material 3-Pyridineacetic acid hydrochloride
CAS 6419-36-9
Molecular Formula C7H7NO2·HCl
Formula Weight 173.6
MDL Number MFCD00012819
Beilstein 3696630
Merck Index 14,7971
Synonym 3-Pyridylacetic acid hydrochloride
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAA1559906

  SDS
alfa aesar™
A1559906
5g Each for $114.66

AAA1559914

  SDS
alfa aesar™
A1559914
25g Each for $354.09

AAA1559922

  SDS
alfa aesar™
A1559922
100g Each for $1,090.89
Description & Specifications

Specifications

Formula Weight 173.6
Chemical Name or Material 3-Pyridineacetic acid hydrochloride
Assay ≥98%
Molecular Formula C7H7NO2·HCl
MDL Number MFCD00012819
CAS 6419-36-9
Beilstein 3696630
Merck Index 14,7971
Synonym 3-Pyridylacetic acid hydrochloride
Health Hazard 1 Warning
Health Hazard 2 GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Health Hazard 3 GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
Melting Point 158° to 162°C
Quantity 5g
Safety and Handling

Warning

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.