missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ 3-Pyridineacetic acid hydrochloride, 98+%

Manufacturer:  Alfa Aesar™ A1559922

 View more versions of this product

Catalog No. AAA1559922



Specifications

Specifications

3-Pyridineacetic acid hydrochloride
100g
C7H8ClNO2
3696630
3-pyridylacetic acid hydrochloride, 2-pyridin-3-yl acetic acid hydrochloride, pyridine-3-acetic acid hydrochloride, 3-pyridineacetic acid hydrochloride, 3-pyridylacetic acid hcl, pyridin-3-ylacetic acid hydrochloride, 3-pyridineacetic acid hcl, 3-pyridinylacetic acid hcl, pyridine-3-acetic acid hcl, 2-3-pyridinyl acetic acid hydrochloride
C1=CC(=CN=C1)CC(=O)O.Cl
173.596
173.6
158°C to 162°C
6419-36-9
MFCD00012819
14,7971
XVCCOEWNFXXUEV-UHFFFAOYSA-N
2-pyridin-3-ylacetic acid;hydrochloride
2723724
≥98%

Chemical Identifiers

6419-36-9
173.596
XVCCOEWNFXXUEV-UHFFFAOYSA-N
2723724
C1=CC(=CN=C1)CC(=O)O.Cl
C7H8ClNO2
MFCD00012819
3-pyridylacetic acid hydrochloride, 2-pyridin-3-yl acetic acid hydrochloride, pyridine-3-acetic acid hydrochloride, 3-pyridineacetic acid hydrochloride, 3-pyridylacetic acid hcl, pyridin-3-ylacetic acid hydrochloride, 3-pyridineacetic acid hcl, 3-pyridinylacetic acid hcl, pyridine-3-acetic acid hcl, 2-3-pyridinyl acetic acid hydrochloride
2-pyridin-3-ylacetic acid;hydrochloride
Safety and Handling

Safety and Handling

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Warning

Documents