Please login to your online account to display your discounted pricing

3-Pyridin-3-ylaniline, 97%, Maybridge


Quantity 5g
Packaging Amber Glass Bottle

Manufacturer: fisher scientific company  CC39814DE

 View more versions of this product

Catalog No. CC39814DE

 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description & Specifications

Chemical Identifiers

Formula Weight 170.21
Quantity 5g
Assay 97%
Chemical Name or Material 3-Pyridin-3-ylaniline
Packaging Amber Glass Bottle
Assay Percent Range 97%
Molecular Formula C11H10N2
CAS 57976-57-5

Specifications

Chemical Name or Material 3-Pyridin-3-ylaniline
Assay 97%
Assay Percent Range 97%
Molecular Formula C11H10N2
Formula Weight 170.21