missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

3-Pyrid-4-ylbenzoic acid, 97%, Maybridge

Chemical Identifiers

Assay Percent Range 95%
Molecular Formula C12H9NO2
Formula Weight 199.21
Chemical Name or Material 3-Pyrid-4-ylbenzoic acid
Assay 97%
View More Specs


Products ${productFamilyLength}
Catalog Number Mfr. No. Packaging Quantity Price Quantity  
Catalog Number Mfr. No. Packaging Quantity Price Quantity  
CC35601DE Fisher Scientific Company
CC35601DE
Amber glass bottle 5g N/A N/A  
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
CC35601DA SDS Fisher Scientific Company
CC35601DA
Amber glass bottle 1g N/A N/A  
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
CC35601CB SDS Fisher Scientific Company
CC35601CB
250mg N/A N/A  
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Specifications Description & Specifications

Chemical Identifiers

95%
199.21
97%
C12H9NO2
3-Pyrid-4-ylbenzoic acid

Specifications

5g
Amber glass bottle
C12H9NO2
3-Pyrid-4-ylbenzoic acid
4385-78-8
95%
199.21
97%

Safety and Handling

Warning