Please login to your online account to display your discounted pricing

Alfa Aesar™ 3-Propionylpyridine, 98+%

$85.43 - $311.91

Chemical Identifiers

Chemical Name or Material 3-Propionylpyridine
CAS 1570-48-5
Molecular Formula C8H9NO
Formula Weight 135.17
MDL Number MFCD00023589
Beilstein 111365
Synonym Ethyl 3-pyridyl ketone
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAL0971103

  SDS
alfa aesar™
L0971103
1g Each for $85.43

AAL0971106

  SDS
alfa aesar™
L0971106
5g Each for $311.91
Description & Specifications

Specifications

Formula Weight 135.17
Health Hazard 3 GHS P Statement
P280-P305+P351+P338-P362-P321-P332+P313-P337+P313
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Take off contaminated clothing and wash before reuse.
Specific treatment (see label).
If skin irritation occurs: Get medical advice/attention.
If eye irritation persists: Get medical advice/attention.
Molecular Formula C8H9NO
Chemical Name or Material 3-Propionylpyridine
MDL Number MFCD00023589
CAS 1570-48-5
Health Hazard 2 GHS H Statement
H315-H319
Causes skin irritation.
Causes serious eye irritation.
Assay ≥98%
Boiling Point 109° to 112°C (10mmHg)
Synonym Ethyl 3-pyridyl ketone
Beilstein 111365
Quantity 1g
Health Hazard 1 Warning
Safety and Handling

Warning

GHS P Statement
P280-P305+P351+P338-P362-P321-P332+P313-P337+P313
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Take off contaminated clothing and wash before reuse.
Specific treatment (see label).
If skin irritation occurs: Get medical advice/attention.
If eye irritation persists: Get medical advice/attention.

GHS H Statement
H315-H319
Causes skin irritation.
Causes serious eye irritation.