missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ 3-Propionylpyridine, 98+%

Manufacturer:  Alfa Aesar™ L0971106

 View more versions of this product

Catalog No. AAL0971106



Specifications

Specifications

3-Propionylpyridine
5g
C8H9NO
111365
VDNKJMUNLKAGAM-UHFFFAOYSA-N
1-pyridin-3-ylpropan-1-one
74074
≥98%
109°C to 112°C (10mmHg)
1570-48-5
MFCD00023589
3-propionylpyridine, 1-3-pyridyl propan-1-one, ethyl 3-pyridyl ketone, 1-pyridin-3-yl propan-1-one, 3-pyridyl ethyl ketone, 1-3-pyridinyl-1-propanone, 3-propionyl pyridine, 1-pyridin-3-yl-propan-1-one, ethyl pyridin-3-yl ketone, 1-propanone, 1-3-pyridinyl
CCC(=O)C1=CN=CC=C1
135.166
135.17

Chemical Identifiers

1570-48-5
135.166
VDNKJMUNLKAGAM-UHFFFAOYSA-N
74074
CCC(=O)C1=CN=CC=C1
C8H9NO
MFCD00023589
3-propionylpyridine, 1-3-pyridyl propan-1-one, ethyl 3-pyridyl ketone, 1-pyridin-3-yl propan-1-one, 3-pyridyl ethyl ketone, 1-3-pyridinyl-1-propanone, 3-propionyl pyridine, 1-pyridin-3-yl-propan-1-one, ethyl pyridin-3-yl ketone, 1-propanone, 1-3-pyridinyl
1-pyridin-3-ylpropan-1-one
Safety and Handling

Safety and Handling

GHS H Statement
H315-H319
Causes skin irritation.
Causes serious eye irritation.

GHS P Statement
P280-P305+P351+P338-P362-P321-P332+P313-P337+P313
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Take off contaminated clothing and wash before reuse.
Specific treatment (see label).
If skin irritation occurs: Get medical advice/attention.
If eye irritation persists: Get medical advice/attention.

Warning

Documents