Please login to your online account to display your discounted pricing

(3-Piperidinophenyl)methylamine, 95%, Maybridge

Chemical Identifiers

Chemical Name or Material (3-Piperidinophenyl)methylamine
Assay 95%
Assay Percent Range 95%
Molecular Formula C12H18N2
Formula Weight 190.29
View More Specs
Products
Catalog Number Mfr. No. Packaging Quantity Price Quantity    

CC34013CB

 
fisher scientific company
CC34013CB
Amber Glass Bottle 250mg CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CC34013DA

 
fisher scientific company
CC34013DA
Amber Glass Bottle 1g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CC34013DE

 
fisher scientific company
CC34013DE
Amber Glass Bottle 5g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Formula Weight 190.29
Quantity 250mg
Assay 95%
Assay Percent Range 95%
Molecular Formula C12H18N2
Chemical Name or Material (3-Piperidinophenyl)methylamine
CAS 175696-71-6
Packaging Amber Glass Bottle