Please login to your online account to display your discounted pricing

(3-Piperidinophenyl)methylamine, 95%, Maybridge


Quantity 5g
Packaging Amber Glass Bottle

Manufacturer: fisher scientific company  CC34013DE

 View more versions of this product

Catalog No. CC34013DE

 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description & Specifications

Chemical Identifiers

Formula Weight 190.29
Quantity 5g
Assay 95%
Chemical Name or Material (3-Piperidinophenyl)methylamine
Molecular Formula C12H18N2
Packaging Amber Glass Bottle
Assay Percent Range 95%
CAS 175696-71-6

Specifications

Chemical Name or Material (3-Piperidinophenyl)methylamine
Assay 95%
Assay Percent Range 95%
Molecular Formula C12H18N2
Formula Weight 190.29