Learn More
3-Piperidinemethanol, 96%, Thermo Scientific Chemicals
$69.33 - $242.08
Chemical Identifiers
CAS | 4606-65-9 |
---|---|
Molecular Formula | C6H13NO |
Molecular Weight (g/mol) | 115.176 |
MDL Number | MFCD00005997 |
InChI Key | VUNPWIPIOOMCPT-UHFFFAOYSA-N |
Synonym | 3-piperidinemethanol, 3-hydroxymethyl piperidine, piperidin-3-yl-methanol, 3-hydroxymethylpiperidine, 3-piperidinylmethanol, piperidine-3-methanol, 3-piperidine methanol, 3-piperidylmethanol, 3-piperidinyl methanol, piperidin-3-yl methanol |
PubChem CID | 107308 |
IUPAC Name | piperidin-3-ylmethanol |
SMILES | C1CC(CNC1)CO |
Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
AAL0426406
|
Thermo Scientific Chemicals
L0426406 |
5 g |
N/A
|
|
|||||
AAL0426414
|
Thermo Scientific Chemicals
L0426414 |
25 g |
N/A
|
|
|||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
4606-65-9 | |
115.176 | |
VUNPWIPIOOMCPT-UHFFFAOYSA-N | |
107308 | |
C1CC(CNC1)CO |
C6H13NO | |
MFCD00005997 | |
3-piperidinemethanol, 3-hydroxymethyl piperidine, piperidin-3-yl-methanol, 3-hydroxymethylpiperidine, 3-piperidinylmethanol, piperidine-3-methanol, 3-piperidine methanol, 3-piperidylmethanol, 3-piperidinyl methanol, piperidin-3-yl methanol | |
piperidin-3-ylmethanol |
Specifications
4606-65-9 | |
1.025 | |
>110°C (230°F) | |
1.494 | |
5 g | |
103058 | |
3-piperidinemethanol, 3-hydroxymethyl piperidine, piperidin-3-yl-methanol, 3-hydroxymethylpiperidine, 3-piperidinylmethanol, piperidine-3-methanol, 3-piperidine methanol, 3-piperidylmethanol, 3-piperidinyl methanol, piperidin-3-yl methanol | |
C1CC(CNC1)CO | |
115.176 | |
115.18 | |
3-Piperidinemethanol |
58°C to 62°C | |
236°C | |
C6H13NO | |
MFCD00005997 | |
UN3259 | |
Hygroscopic | |
VUNPWIPIOOMCPT-UHFFFAOYSA-N | |
piperidin-3-ylmethanol | |
107308 | |
96% |
Safety and Handling
GHS H Statement
H314-H318
Causes severe skin burns and eye damage.
Causes serious eye damage.
P260-P264b-P271-P280-P301+P330+P331-P303+P361+P353-P304+P340-P305+P351+P338-P310-P363-P501c
H314-H335
DOTInformation : Hazard Class: 8; Packaging Group: II
EINECSNumber : 225-013-0
TSCA : No
Recommended Storage : Ambient temperatures
RUO – Research Use Only