missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

3-Piperidinecarboxylic acid, 97%

Manufacturer:  Thermo Scientific AC19919DA

 View more versions of this product

Catalog No. AC19919DA

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Specifications

Specifications

3-Piperidinecarboxylic acid
Amber Glass Bottle
1g
498-95-3
100.0
C6H11NO2
+/--3-piperidine carboxylic acid
C1CC(CNC1)C(=O)O
129.159
CHEBI:116931
Powder
Authentic
Beige-Yellow to White
Functional Group amine (alkyl)
     carboxylic acid
97.5
97%
MFCD00005992
XJLSEXAGTJCILF-UHFFFAOYSA-N
piperidine-3-carboxylic acid
4498
129.16
≥97.5%

Chemical Identifiers

498-95-3
129.159
XJLSEXAGTJCILF-UHFFFAOYSA-N
4498
piperidine-3-carboxylic acid
C6H11NO2
MFCD00005992
+/--3-piperidine carboxylic acid
CHEBI:116931
C1CC(CNC1)C(=O)O
Safety and Handling

Safety and Handling

GHS H Statement:
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

SDS Documents