missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

3-(Piperazin-1-yl)-1,2-benzisoxazole, 95%, ACROS Organics™

Manufacturer:  Thermo Fisher Scientific AC446700010

 View more versions of this product

Catalog No. AC446700010

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Specifications

Specifications

3-(Piperazin-1-yl)-1, 2-benzisoxazole
69.0°C to 72.0°C
87691-89-2
C11H13N3O
ZDFQBFVFCPABKQ-UHFFFAOYSA-N
3-piperazin-1-yl-1,2-benzoxazole
13232475
95%
Glass Bottle
1g
94% min. (GC)
3-1-piperazino-1,2-benzisoxazole, 3-1-piperazinyl-1,2-benzisoxazole, 3-1-piperazinyl-1,2-benzoxazole, 3-piperazin-1-yl benzo d isoxazole, 3-piperazin-1-yl-1,2-benzisoxazole, 3-piperazin-1-yl-1,2-benzoxazole, 3-piperazin-1-yl-benzo d isoxazole, 3-piperazinylbenzo d isoxazole, 4 1,2-benzisoxazol-3-yl piperazine
C1CN(CCN1)C2=NOC3=CC=CC=C32
203.24
203.24

Chemical Identifiers

87691-89-2
203.24
3-1-piperazino-1,2-benzisoxazole, 3-1-piperazinyl-1,2-benzisoxazole, 3-1-piperazinyl-1,2-benzoxazole, 3-piperazin-1-yl benzo d isoxazole, 3-piperazin-1-yl-1,2-benzisoxazole, 3-piperazin-1-yl-1,2-benzoxazole, 3-piperazin-1-yl-benzo d isoxazole, 3-piperazinylbenzo d isoxazole, 4 1,2-benzisoxazol-3-yl piperazine
3-piperazin-1-yl-1,2-benzoxazole
C11H13N3O
ZDFQBFVFCPABKQ-UHFFFAOYSA-N
13232475
C1CN(CCN1)C2=NOC3=CC=CC=C32
Safety and Handling

Safety and Handling

GHS H Statement
Causes skin irritation.
Causes serious eye irritation.

GHS P Statement
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses,if present

GHS Signal Word: Warning

SDS Documents