Please login to your online account to display your discounted pricing

3-(Piperazin-1-yl)-1,2-benzisoxazole, 95%, ACROS Organics™

Chemical Identifiers

Chemical Name or Material 3-(Piperazin-1-yl)-1, 2-benzisoxazole
Assay Percent Range 95%
Molecular Formula C11H13N3O
Formula Weight 203.24
Synonym 1-(Benzisoxazol-3-yl)piperazine
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AC446700010

 
thermo fisher scientific
AC446700010
1g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AC446700050

 
thermo fisher scientific
AC446700050
5g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Chemical Name or Material 3-(Piperazin-1-yl)-1, 2-benzisoxazole
Quantity 1g
CAS 87691-89-2
Assay Percent Range 95%
Molecular Formula C11H13N3O
Formula Weight 203.24
Synonym 1-(Benzisoxazol-3-yl)piperazine