Please login to your online account to display your discounted pricing

Alfa Aesar™ 3-Picoline N-oxide, 98%

$63.57 - $200.11

Chemical Identifiers

Chemical Name or Material 3-Picoline N-oxide
CAS 1003-73-2
Molecular Formula C6H7NO
Formula Weight 109.13
MDL Number MFCD00006203
Beilstein 106327
Synonym 3-Methylpyridine N-oxide
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAA1257822

  SDS
alfa aesar™
A1257822
100g Each for $63.57

AAA1257836

  SDS
alfa aesar™
A1257836
500g Each for $200.11
Description & Specifications

Specifications

Formula Weight 109.13
Chemical Name or Material 3-Picoline N-oxide
TSCA TSCA
Assay 98%
Molecular Formula C6H7NO
MDL Number MFCD00006203
CAS 1003-73-2
Beilstein 106327
Synonym 3-Methylpyridine N-oxide
Health Hazard 1 Warning
Health Hazard 2 GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Health Hazard 3 GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
RTECS Number UT5799000
Flash Point 155°C (311°F)
Boiling Point 150°C (15mmHg)
Melting Point 36° to 40°C
Quantity 100g
Safety and Handling

Warning

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.