missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

3-Phthalimidopropionic Acid, TCI America™

Manufacturer:  TCI America P210125G

 View more versions of this product

Catalog No. P210125G



Specifications

Specifications

3-Phthalimidopropionic Acid
153°C
3339-73-9
MFCD00023096
DXXHRZUOTPMGEH-UHFFFAOYSA-N
3-(1,3-dioxoisoindol-2-yl)propanoic acid
76859
Crystal-Powder at 20°C
White
25g
C11H9NO4
3-phthalimidopropionic acid, 3-1,3-dioxoisoindolin-2-yl propanoic acid, 2h-isoindole-2-propanoic acid, 1,3-dihydro-1,3-dioxo, 3-1,3-dioxo-1,3-dihydro-2h-isoindol-2-yl propanoic acid, 3-1,3-dioxo-2,3-dihydro-1h-isoindol-2-yl propanoic acid, pht-beta-ala-oh, b-phthalimidopropionic acid, 3-phthalimidopropanoic acid, n-phthaloyl-beta-alanine, phthalyl-.beta.-alanine
C1=CC=C2C(=C1)C(=O)N(C2=O)CCC(=O)O
219.196
219.20
≥98.0% (HPLC,T)

Chemical Identifiers

3339-73-9
219.196
DXXHRZUOTPMGEH-UHFFFAOYSA-N
76859
C1=CC=C2C(=C1)C(=O)N(C2=O)CCC(=O)O
C11H9NO4
MFCD00023096
3-phthalimidopropionic acid, 3-1,3-dioxoisoindolin-2-yl propanoic acid, 2h-isoindole-2-propanoic acid, 1,3-dihydro-1,3-dioxo, 3-1,3-dioxo-1,3-dihydro-2h-isoindol-2-yl propanoic acid, 3-1,3-dioxo-2,3-dihydro-1h-isoindol-2-yl propanoic acid, pht-beta-ala-oh, b-phthalimidopropionic acid, 3-phthalimidopropanoic acid, n-phthaloyl-beta-alanine, phthalyl-.beta.-alanine
3-(1,3-dioxoisoindol-2-yl)propanoic acid
Safety and Handling

Safety and Handling

TSCA : No

Documents