Learn More
3-Phthalimidopropionic acid, 98%, Thermo Scientific Chemicals
$62.19 - $190.95
Chemical Identifiers
CAS | 3339-73-9 |
---|---|
Molecular Formula | C11H9NO4 |
Molecular Weight (g/mol) | 219.196 |
MDL Number | MFCD00023096 |
InChI Key | DXXHRZUOTPMGEH-UHFFFAOYSA-N |
Synonym | 3-phthalimidopropionic acid, 3-1,3-dioxoisoindolin-2-yl propanoic acid, 2h-isoindole-2-propanoic acid, 1,3-dihydro-1,3-dioxo, 3-1,3-dioxo-1,3-dihydro-2h-isoindol-2-yl propanoic acid, 3-1,3-dioxo-2,3-dihydro-1h-isoindol-2-yl propanoic acid, pht-beta-ala-oh, b-phthalimidopropionic acid, 3-phthalimidopropanoic acid, n-phthaloyl-beta-alanine, phthalyl-.beta.-alanine |
PubChem CID | 76859 |
IUPAC Name | 3-(1,3-dioxoisoindol-2-yl)propanoic acid |
SMILES | C1=CC=C2C(=C1)C(=O)N(C2=O)CCC(=O)O |
Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
AAL1353503
|
Thermo Scientific Chemicals
L1353503 |
1 g |
Each for $62.19
|
|
|||||
AAL1353506
|
Thermo Scientific Chemicals
L1353506 |
5 g |
Each for $190.95
|
|
|||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
3339-73-9 | |
219.196 | |
DXXHRZUOTPMGEH-UHFFFAOYSA-N | |
76859 | |
C1=CC=C2C(=C1)C(=O)N(C2=O)CCC(=O)O |
C11H9NO4 | |
MFCD00023096 | |
3-phthalimidopropionic acid, 3-1,3-dioxoisoindolin-2-yl propanoic acid, 2h-isoindole-2-propanoic acid, 1,3-dihydro-1,3-dioxo, 3-1,3-dioxo-1,3-dihydro-2h-isoindol-2-yl propanoic acid, 3-1,3-dioxo-2,3-dihydro-1h-isoindol-2-yl propanoic acid, pht-beta-ala-oh, b-phthalimidopropionic acid, 3-phthalimidopropanoic acid, n-phthaloyl-beta-alanine, phthalyl-.beta.-alanine | |
3-(1,3-dioxoisoindol-2-yl)propanoic acid |
Specifications
3339-73-9 | |
C11H9NO4 | |
1 g | |
3-phthalimidopropionic acid, 3-1,3-dioxoisoindolin-2-yl propanoic acid, 2h-isoindole-2-propanoic acid, 1,3-dihydro-1,3-dioxo, 3-1,3-dioxo-1,3-dihydro-2h-isoindol-2-yl propanoic acid, 3-1,3-dioxo-2,3-dihydro-1h-isoindol-2-yl propanoic acid, pht-beta-ala-oh, b-phthalimidopropionic acid, 3-phthalimidopropanoic acid, n-phthaloyl-beta-alanine, phthalyl-.beta.-alanine | |
C1=CC=C2C(=C1)C(=O)N(C2=O)CCC(=O)O | |
219.196 | |
219.2 | |
3-Phthalimidopropionic acid |
151°C to 153°C | |
MFCD00023096 | |
15570 | |
DXXHRZUOTPMGEH-UHFFFAOYSA-N | |
3-(1,3-dioxoisoindol-2-yl)propanoic acid | |
76859 | |
98% |
Safety and Handling
GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P312-P330-P332+P313-P362-P501c
H302-H315-H319-H335
EINECSNumber : 222-083-4
TSCA : No
Recommended Storage : Ambient temperatures
RUO – Research Use Only