Please login to your online account to display your discounted pricing

3-Phthalimidopropionic acid, +97% , ACROS Organics™


Quantity 5g
Packaging Glass bottle

Manufacturer: thermo fisher scientific  AC391000050

 View more versions of this product

Catalog No. AC391000050

 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description & Specifications

Chemical Identifiers

Assay ≥97.0%
Physical Form Crystalline Powder
Formula Weight 219.2
Quantity 5g
Color White
Melting Point 149° to 153°C
CAS Min % 97.0
CAS Max % 100.0
Chemical Name or Material 3-Phthalimidopropionic acid
Packaging Glass bottle
Assay Percent Range 97+%
MDL Number MFCD00023096
CAS 3339-73-9
Health Hazard 3 GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
Molecular Formula C11H9NO4
Synonym 3-(1, 3-Dioxo-2, 3-dihydro-1H-isoindol-2-yl)propanoic acid
Health Hazard 2 GHS H Statement:
May cause respiratory irritation.
Causes skin irritation.
Harmful if swallowed.
Causes serious eye irritation.
Infrared Spectrum Authentic

Specifications

Chemical Name or Material 3-Phthalimidopropionic acid
CAS 3339-73-9
CAS Min % 97.0
CAS Max % 100.0
Assay ≥97.0%
Assay Percent Range 97+%
Molecular Formula C11H9NO4
Formula Weight 219.2
MDL Number MFCD00023096
Synonym 3-(1, 3-Dioxo-2, 3-dihydro-1H-isoindol-2-yl)propanoic acid
Accessories

GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS H Statement:
May cause respiratory irritation.
Causes skin irritation.
Harmful if swallowed.
Causes serious eye irritation.

SDS & Safety Documents