Please login to your online account to display your discounted pricing

Alfa Aesar™ 3-Phosphonopropionic acid, 94%

Chemical Identifiers

Chemical Name or Material 3-Phosphonopropionic acid
CAS 5962-42-5
Molecular Formula C3H7O5P
Formula Weight 154.06
MDL Number MFCD00002770
UN Number UN3261
Beilstein 1768923
Synonym 2-Carboxyethylphosphonic acid
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAL1040206

  SDS
alfa aesar™
L1040206
5g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Formula Weight 154.06
MDL Number MFCD00002770
Assay 94%
Chemical Name or Material 3-Phosphonopropionic acid
Molecular Formula C3H7O5P
Beilstein 1768923
DOT Information Transport Hazard Class: 8; Packing Group: III; Proper Shipping Name: CORROSIVE SOLID, ACIDIC, ORGANIC, N.O.S.
Melting Point 162° to 166°C
Odor Odorless
Health Hazard 1 Danger
Health Hazard 3 GHS P Statement
P280-P305+P351+P338-P309-P310
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF exposed or if you feel unwell: Immediately call a POISON CENTER or doctor/physician.
TSCA TSCA
Synonym 2-Carboxyethylphosphonic acid
RTECS Number UA2558437
Health Hazard 2 GHS H Statement
H314-H318
Causes severe skin burns and eye damage.
Causes serious eye damage.
Quantity 5g
UN Number UN3261
CAS 5962-42-5
Safety and Handling

Danger

GHS P Statement
P280-P305+P351+P338-P309-P310
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF exposed or if you feel unwell: Immediately call a POISON CENTER or doctor/physician.

GHS H Statement
H314-H318
Causes severe skin burns and eye damage.
Causes serious eye damage.