Please login to your online account to display your discounted pricing

Alfa Aesar™ 3-(Phenylsulfonyl)thiophene, 98%

$50.18 - $152.39

Chemical Identifiers

Chemical Name or Material 3-(Phenylsulfonyl)thiophene
CAS 16718-05-1
Molecular Formula C10H8O2S2
Formula Weight 224.3
MDL Number MFCD00052136
Synonym 3-(Benzenesulfonyl)thiophene; Phenyl 3-thienyl ketone
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAA1336103

 
alfa aesar™
A1336103
1g Each for $152.39

AAA1336101

 
alfa aesar™
A1336101
0.25g Each for $50.18
Description & Specifications

Specifications

Formula Weight 224.3
Chemical Name or Material 3-(Phenylsulfonyl)thiophene
Assay 98%
Molecular Formula C10H8O2S2
MDL Number MFCD00052136
CAS 16718-05-1
Synonym 3-(Benzenesulfonyl)thiophene; Phenyl 3-thienyl ketone
Melting Point 115° to 117°C
Quantity 1g