missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ 3-Phenylsulfonamidopyridine-5-boronic acid pinacol ester, 96%

Manufacturer:  Alfa Aesar™ H54339MD

 View more versions of this product

Catalog No. AAH54339MD



Description

Description

Specifications

Specifications

3-Phenylsulfonamidopyridine-5-boronic acid pinacol ester
1083326-28-6
MFCD13190589
UXJVHVXONVGHIL-UHFFFAOYSA-N
N-[5-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)pyridin-3-yl]benzenesulfonamide
52936632
96%
250mg
C17H21BN2O4S
3-phenylsulfonamidopyridine-5-boronic acid pinacol ester, n-5-4,4,5,5-tetramethyl-1,3,2 dioxaborolan-2-yl pyridin-3-yl benzenesulfonamide, n-5-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl pyridin-3-yl benzenesulfonamide, n-5-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl-3-pyridinyl benzenesulfonamide, n-5-tetramethyl-1,3,2-dioxaborolan-2-yl pyridin-3-yl benzenesulfonamide
B1(OC(C(O1)(C)C)(C)C)C2=CC(=CN=C2)NS(=O)(=O)C3=CC=CC=C3
360.235
360.24

Chemical Identifiers

1083326-28-6
360.235
UXJVHVXONVGHIL-UHFFFAOYSA-N
52936632
B1(OC(C(O1)(C)C)(C)C)C2=CC(=CN=C2)NS(=O)(=O)C3=CC=CC=C3
C17H21BN2O4S
MFCD13190589
3-phenylsulfonamidopyridine-5-boronic acid pinacol ester, n-5-4,4,5,5-tetramethyl-1,3,2 dioxaborolan-2-yl pyridin-3-yl benzenesulfonamide, n-5-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl pyridin-3-yl benzenesulfonamide, n-5-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl-3-pyridinyl benzenesulfonamide, n-5-tetramethyl-1,3,2-dioxaborolan-2-yl pyridin-3-yl benzenesulfonamide
N-[5-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)pyridin-3-yl]benzenesulfonamide
Safety and Handling

Safety and Handling

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c

H315-H319-H335

TSCA : No

Recommended Storage : Ambient temperatures

SDS
Documents