missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ 3-Phenylsulfonamidopyridine-5-boronic acid pinacol ester, 96%

Manufacturer:  Alfa Aesar™ H5433903

 View more versions of this product

Catalog No. AAH5433903



Specifications

Specifications

3-Phenylsulfonamidopyridine-5-boronic acid pinacol ester
1083326-28-6
MFCD13190589
UXJVHVXONVGHIL-UHFFFAOYSA-N
N-[5-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)pyridin-3-yl]benzenesulfonamide
52936632
96%
1g
C17H21BN2O4S
3-phenylsulfonamidopyridine-5-boronic acid pinacol ester
B1(OC(C(O1)(C)C)(C)C)C2=CC(=CN=C2)NS(=O)(=O)C3=CC=CC=C3
360.235
360.24

Chemical Identifiers

1083326-28-6
360.235
UXJVHVXONVGHIL-UHFFFAOYSA-N
52936632
B1(OC(C(O1)(C)C)(C)C)C2=CC(=CN=C2)NS(=O)(=O)C3=CC=CC=C3
C17H21BN2O4S
MFCD13190589
3-phenylsulfonamidopyridine-5-boronic acid pinacol ester
N-[5-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)pyridin-3-yl]benzenesulfonamide
Safety and Handling

Safety and Handling

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Warning

Documents