missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ 3-Phenylpropyl isothiocyanate, 98%

Manufacturer:  Alfa Aesar™ L1093106

 View more versions of this product

Catalog No. AAL1093106



Specifications

Specifications

3-Phenylpropyl isothiocyanate
156°C to 160°C (12mmHg)
2627-27-2
MFCD00041134
2209321
GRUOGLPIAPZLHJ-UHFFFAOYSA-N
3-isothiocyanatopropylbenzene
75815
98%
Irritating
5g
C10H11NS
UN2922
3-phenylpropyl isothiocyanate, 3-isothiocyanatopropyl benzene, phenylpropyl isothiocyanate, ppitc, benzene, 3-isothiocyanatopropyl, unii-1s20ia5ggo, ccris 3147, 3phenylpropylisothiocyanate, 3-phenylpropylisothiocyanate, 3-phenylpropyl isothiocynate
C1=CC=C(C=C1)CCCN=C=S
177.265
177.27

Chemical Identifiers

2627-27-2
177.265
GRUOGLPIAPZLHJ-UHFFFAOYSA-N
75815
C1=CC=C(C=C1)CCCN=C=S
C10H11NS
MFCD00041134
3-phenylpropyl isothiocyanate, 3-isothiocyanatopropyl benzene, phenylpropyl isothiocyanate, ppitc, benzene, 3-isothiocyanatopropyl, unii-1s20ia5ggo, ccris 3147, 3phenylpropylisothiocyanate, 3-phenylpropylisothiocyanate, 3-phenylpropyl isothiocynate
3-isothiocyanatopropylbenzene
Safety and Handling

Safety and Handling

GHS H Statement
H331-H314-H318-H302-H312
Toxic if inhaled.
Causes severe skin burns and eye damage.
Causes serious eye damage.
Harmful if swallowed.
Harmful in contact with skin.

GHS P Statement
P260-P303+P361+P353-P305+P351+P338-P301+P330+P331-P405-P501a
Do not breathe dust/fume/gas/mist/vapors/spray.
IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing. Rinse skin with water/shower.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF SWALLOWED: Rinse mouth. Do NOT induce vomiting.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Danger

DOTInformation : Transport Hazard Class: 8; Packing Group: III; Proper Shipping Name: CORROSIVE LIQUIDS, TOXIC, N.O.S.

Documents