missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ 3-Phenylpropionic acid, 99%

Manufacturer:  Alfa Aesar™ A1490836

 View more versions of this product

Catalog No. AAA1490836



Description

Description

Applications
Hydrocinnamic acid is inhibitor of carboxypeptidase. Hydrocinnamic acid was used in the synthesis of three medium-chain acyl-Coenzyme A′s i.e. 3-phenylpropionyl-CoA from mixed anhydrides of fatty acids.

Solubility
Soluble in water

Notes
Store away from strong oxidizing agents. Keep container tightly closed. Store in cool, dry conditions in well sealed containers.
Specifications

Specifications

3-Phenylpropionic acid
280°C
500g
C9H10O2
907515
hydrocinnamic acid, 3-phenylpropionic acid, benzylacetic acid, benzenepropanoic acid, dihydrocinnamic acid, benzenepropionic acid, phenylpropanoic acid, 3-phenyl-propionic acid, phenylpropanoate, beta-phenylpropionic acid
C1=CC=C(C=C1)CCC(=O)O
150.177
CHEBI:28631
99%
>110°C (230°F)
47°C to 50°C
501-52-0
MFCD00002771
14,4784
XMIIGOLPHOKFCH-UHFFFAOYSA-N
3-phenylpropanoic acid
107
150.18

Chemical Identifiers

501-52-0
150.177
XMIIGOLPHOKFCH-UHFFFAOYSA-N
107
3-phenylpropanoic acid
C9H10O2
MFCD00002771
hydrocinnamic acid, 3-phenylpropionic acid, benzylacetic acid, benzenepropanoic acid, dihydrocinnamic acid, benzenepropionic acid, phenylpropanoic acid, 3-phenyl-propionic acid, phenylpropanoate, beta-phenylpropionic acid
CHEBI:28631
C1=CC=C(C=C1)CCC(=O)O
Safety and Handling

Safety and Handling

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Warning

RTECSNumber : DA8600000

TSCA : TSCA

Documents