missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
3-Phenylpropionaldehyde, 95%, ACROS Organics™
Manufacturer: Thermo Fisher Scientific AC240515000
Specifications
3-Phenylpropionaldehyde | |
95°C | |
Glass bottle | |
1.01 | |
224°C to 226°C | |
-42°C | |
104-53-0 | |
100.0 | |
C9H10O | |
MFCD00007021 | |
Solubility in water: slightly soluble. Other solubilities: soluble in 50% alcohol | |
C1=CC=C(C=C1)CCC=O | |
134.18 | |
134.18 | |
95% |
1.0100g/mL | |
Authentic | |
1.5200 to 1.525 | |
2.3 mPa.s (25°C) | |
Colorless to Yellow | |
500g | |
94.0 | |
94% min. (GC) | |
C6H5CH2CH2CHO | |
3-phenyl-1-propanal, 3-phenylpropionaldehyde, 3-phenylpropyl aldehyde, 3-phenylpropylaldehyde, benzenepropanal, benzylacetaldehyde, dihydrocinnamaldehyde, hydrocinnamaldehyde, hydrocinnamic aldehyde, propanal, phenyl | |
YGCZTXZTJXYWCO-UHFFFAOYSA-N | |
3-phenylpropanal | |
7707 | |
Liquid |
Chemical Identifiers
104-53-0 | |
134.18 | |
YGCZTXZTJXYWCO-UHFFFAOYSA-N | |
7707 | |
C1=CC=C(C=C1)CCC=O |
C9H10O | |
MFCD00007021 | |
3-phenyl-1-propanal, 3-phenylpropionaldehyde, 3-phenylpropyl aldehyde, 3-phenylpropylaldehyde, benzenepropanal, benzylacetaldehyde, dihydrocinnamaldehyde, hydrocinnamaldehyde, hydrocinnamic aldehyde, propanal, phenyl | |
3-phenylpropanal |
Safety and Handling
GHS H Statement
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses,if present and easy to do.
Continue rinsing.
GHS Signal Word: Warning
EINECSNumber : 203-211-8