Please login to your online account to display your discounted pricing

Alfa Aesar™ 3-Phenylphthalide, 99%

$218.58 - $218.58

Chemical Identifiers

Chemical Name or Material 3-Phenylphthalide
CAS 5398-11-8
Molecular Formula C14H10O2
Formula Weight 210.23
MDL Number MFCD00023099
Beilstein 159678
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAL1618114

 
alfa aesar™
L1618114
25g Each for $218.58
Description & Specifications

Specifications

Formula Weight 210.23
CAS 5398-11-8
Assay 99%
Molecular Formula C14H10O2
MDL Number MFCD00023099
Beilstein 159678
RTECS Number TI3800000
Melting Point 116° to 118°C
Quantity 25g
Chemical Name or Material 3-Phenylphthalide