missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

3-Phenylphenol, 90%, ACROS Organics™

Manufacturer:  Thermo Fisher Scientific AC417660050

 View more versions of this product

Catalog No. AC417660050



Specifications

Specifications

3-Phenylphenol
Glass bottle
Beige-Yellow to White
5g
580-51-8
100.0
C12H10O
1,1'-biphenyl-3-ol, 3-biphenylol, 3-hydroxybiphenyl, 3-hydroxydiphenyl, 3-phenyl phenol, biphenyl-3-ol, m-hydroxybiphenyl, m-hydroxydiphenyl, m-phenylphenol, unii-zu11x47h4o
UBXYXCRCOKCZIT-UHFFFAOYSA-N
3-phenylphenol
11381
170.21
90%
Authentic
300.0°C
72.0°C to 80.0°C
90%
88.0
88% min. (GC)
MFCD00002294
Solubility in water: slightly soluble. Other solubilities: organic solvents: ethanol,diethyl ether,chlorof,benzene,pyridine
C1=CC=C(C=C1)C2=CC(=CC=C2)O
170.21
CHEBI:34338
Crystalline Powder or Chunks

Chemical Identifiers

580-51-8
170.21
UBXYXCRCOKCZIT-UHFFFAOYSA-N
11381
3-phenylphenol
C12H10O
MFCD00002294
1,1'-biphenyl-3-ol, 3-biphenylol, 3-hydroxybiphenyl, 3-hydroxydiphenyl, 3-phenyl phenol, biphenyl-3-ol, m-hydroxybiphenyl, m-hydroxydiphenyl, m-phenylphenol, unii-zu11x47h4o
CHEBI:34338
C1=CC=C(C=C1)C2=CC(=CC=C2)O
Safety and Handling

Safety and Handling

GHS H Statement:
May cause respiratory irritation.
Causes serious eye irritation.
Causes skin irritation.

GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS Signal Word: Warning

TSCA : TSCA

SDS Documents