Learn More
3-Phenylisoxazole-5-carboxylic acid, 97%, Thermo Scientific™
Supplier: Thermo Scientific Chemicals CC64501DA
Specifications
3-Phenylisoxazole-5-carboxylic acid | |
96.5 | |
97% | |
C10H7NO3 | |
1g | |
YGTFDJRCCBKLDM-UHFFFAOYSA-N | |
3-phenyl-1,2-oxazole-5-carboxylic acid | |
737474 | |
97% |
14442-12-7 | |
100.0 | |
Amber Glass Bottle | |
MFCD01936010 | |
3-phenylisoxazole-5-carboxylic acid, 3-phenyl-5-isoxazolecarboxylic acid, 5-isoxazolecarboxylic acid, 3-phenyl, 3-phenyl-isoxazole-5-carboxylic acid, phenylisoxazolecarboxylicacid, 5-carboxy-3-phenylisoxazole, 3-phenylisoxazole-5-carboxylic, 3-phenyl-5-isoxazolecarboxylicacid, 3-phenylisoxazole-5-carboxylicacid, 3-phenyl-5-isoxazole carboxylic acid | |
C1=CC=C(C=C1)C2=NOC(=C2)C(=O)O | |
189.17 | |
189.17 |
Chemical Identifiers
14442-12-7 | |
189.17 | |
YGTFDJRCCBKLDM-UHFFFAOYSA-N | |
737474 | |
C1=CC=C(C=C1)C2=NOC(=C2)C(=O)O |
C10H7NO3 | |
MFCD01936010 | |
3-phenylisoxazole-5-carboxylic acid, 3-phenyl-5-isoxazolecarboxylic acid, 5-isoxazolecarboxylic acid, 3-phenyl, 3-phenyl-isoxazole-5-carboxylic acid, phenylisoxazolecarboxylicacid, 5-carboxy-3-phenylisoxazole, 3-phenylisoxazole-5-carboxylic, 3-phenyl-5-isoxazolecarboxylicacid, 3-phenylisoxazole-5-carboxylicacid, 3-phenyl-5-isoxazole carboxylic acid | |
3-phenyl-1,2-oxazole-5-carboxylic acid |
Safety and Handling
GHS H Statement:
Causes skin irritation.
Harmful if swallowed.
Causes serious eye irritation.
May cause respiratory irritation.
GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
GHS Signal Word: Warning