Please login to your online account to display your discounted pricing

Alfa Aesar™ 3-Phenylindole-2-carboxylic acid, 97%

Chemical Identifiers

Chemical Name or Material 3-Phenylindole-2-carboxylic acid
CAS 6915-67-9
Formula Weight 237.3
MDL Number MFCD00974212
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAH33246MD

 
alfa aesar™
H33246MD
250mg CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AAH3324603

 
alfa aesar™
H3324603
1g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Formula Weight 237.3
Melting Point 195° to 197°C
Chemical Name or Material 3-Phenylindole-2-carboxylic acid
Physical Form Solid
Assay 97%
MDL Number MFCD00974212
CAS 6915-67-9
Health Hazard 1 Warning
Health Hazard 2 GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Health Hazard 3 GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses, if present and easy to do.
Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
Color White to cream
Quantity 250mg

Applications
Reactant for preparation of anticancer agents, derivatives of amino acids and peptides, primary acylureas.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store away from strong oxidizing agents.

Safety and Handling

Warning

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses, if present and easy to do.
Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.