Please login to your online account to display your discounted pricing

Alfa Aesar™ 3-Phenylglutaric acid, 98%

$85.58 - $994.46

Chemical Identifiers

Chemical Name or Material 3-Phenylglutaric acid
CAS 4165-96-2
Molecular Formula C11H12O4
Formula Weight 208.21
MDL Number MFCD00002718
Beilstein 2052171
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAA1457106

  SDS
alfa aesar™
A1457106
5g Each for $85.58

AAA1457114

  SDS
alfa aesar™
A1457114
25g Each for $310.39

AAA1457122

  SDS
alfa aesar™
A1457122
100g Each for $994.46
Description & Specifications

Specifications

Formula Weight 208.21
Molecular Formula C11H12O4
Health Hazard 2 GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Health Hazard 3 GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
Health Hazard 1 Warning
Chemical Name or Material 3-Phenylglutaric acid
Assay 98%
Beilstein 2052171
MDL Number MFCD00002718
Quantity 5g
CAS 4165-96-2
Melting Point 140° to 143°C
Safety and Handling

Warning

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.